ENA428073042 Enamine

5-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)pentanamide

  • MDL number MFCD28064258
  • Cas Number : 4403-43-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177286

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177286
Availability details for
Quantity
  • Availability
  • Add to Cart