ENA428073046 Enamine

6-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)hexanamide

  • MDL number MFCD00266243
  • Cas Number : 141391-34-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177291

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177291
Availability details for
Quantity
  • Availability
  • Add to Cart