ENA428073055 Enamine

1-(2-oxopropyl)cyclobutane-1-carboxylic acid

  • MDL number MFCD28064226
  • Cas Number : 1795306-43-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177350

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177350
Availability details for
Quantity
  • Availability
  • Add to Cart