ENA428073056 Enamine

2-(2-methoxyethyl)oxirane

  • MDL number MFCD22414078
  • Cas Number : 135579-72-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177359

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177359
Availability details for
Quantity
  • Availability
  • Add to Cart