ENA428073064 Enamine

6-aminoquinoline-2-thiol

  • MDL number MFCD12173250
  • Cas Number : 1154583-22-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177412

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177412
Availability details for
Quantity
  • Availability
  • Add to Cart