ENA428073069 Enamine

2-chloro-3-(1,3-dioxolan-2-yl)-6-fluorobenzaldehyde

  • MDL number MFCD27933085
  • Cas Number : 924626-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177446

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177446
Availability details for
Quantity
  • Availability
  • Add to Cart