ENA428073071 Enamine

2-(2-chloro-4-fluorophenyl)-1,3-dioxolane

  • MDL number MFCD06209131
  • Cas Number : 773102-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-177448
Availability details for
Quantity
  • Availability
  • Add to Cart