ENA428073080 Enamine

2-amino-3-(4-fluorophenyl)propan-1-ol hydrochloride

  • MDL number MFCD08454679
  • Cas Number : 1260817-75-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177507

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177507
Availability details for
Quantity
  • Availability
  • Add to Cart