ENA428073082 Enamine

4,6-dichloro-N-(4-methoxyphenyl)pyrimidin-2-amine

  • MDL number MFCD10604277
  • Cas Number : 335649-13-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177521

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177521
Availability details for
Quantity
  • Availability
  • Add to Cart