ENA428073104 Enamine

methyl 3-amino-6-bromo-2-methylbenzoate

  • MDL number MFCD22571479
  • Cas Number : 750586-06-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177554

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177554
Availability details for
Quantity
  • Availability
  • Add to Cart