ENA428073106 Enamine

1-tert-butyl-5-phenyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD10451914
  • Cas Number : 936129-68-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-177567
Availability details for
Quantity
  • Availability
  • Add to Cart