ENA428073113 Enamine

ethyl 2-cyano-4-phenylbutanoate

  • MDL number MFCD24160720
  • Cas Number : 93031-12-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177618

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177618
Availability details for
Quantity
  • Availability
  • Add to Cart