ENA428073115 Enamine

5-amino-2-(trifluoromethyl)benzonitrile

  • MDL number MFCD20694851
  • Cas Number : 354814-19-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-177624
Availability details for
Quantity
  • Availability
  • Add to Cart