ENA428073117 Enamine

1-[4-(trifluoromethyl)phenyl]butane-1,3-dione

  • MDL number MFCD07772941
  • Cas Number : 258346-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177629

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177629
Availability details for
Quantity
  • Availability
  • Add to Cart