ENA428073132 Enamine

5-ethyl-1-(2-methylphenyl)-1H-pyrazol-4-amine

  • MDL number MFCD21773838
  • Cas Number : 1245007-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177681

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177681
Availability details for
Quantity
  • Availability
  • Add to Cart