ENA428073134 Enamine

ethyl 5-(3-cyanophenyl)-1H-1,2,4-triazole-3-carboxylate

  • MDL number MFCD19482168
  • Cas Number : 1284001-05-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177692

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177692
Availability details for
Quantity
  • Availability
  • Add to Cart