ENA428073136 Enamine

1-[(4-fluorophenyl)methyl]-2-oxocyclopentane-1-carbonitrile

  • MDL number MFCD21296170
  • Cas Number : 1493457-80-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177699

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177699
Availability details for
Quantity
  • Availability
  • Add to Cart