ENA428073140 Enamine

2-amino-3-cyclohexylpropanoic acid hydrochloride

  • MDL number MFCD06656846
  • Cas Number : 99065-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177746

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177746
Availability details for
Quantity
  • Availability
  • Add to Cart