ENA428073142 Enamine

methyl 1-cyclopentylpiperidine-2-carboxylate

  • MDL number MFCD23673654
  • Cas Number : 1543455-71-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-177749
Availability details for
Quantity
  • Availability
  • Add to Cart