ENA428073143 Enamine

methyl 2-(2-amino-3-bromophenyl)acetate

  • MDL number MFCD18398641
  • Cas Number : 1261762-84-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-177759

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177759
Availability details for
Quantity
  • Availability
  • Add to Cart