ENA428073146 Enamine

2-(3-chlorophenyl)-1,3,4-oxadiazole

  • MDL number MFCD09881167
  • Cas Number : 5378-33-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177762

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177762
Availability details for
Quantity
  • Availability
  • Add to Cart