ENA428073147 Enamine

tert-butyl 4-amino-3-phenylpiperidine-1-carboxylate

  • MDL number MFCD08751920
  • Cas Number : 632352-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177763

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177763
Availability details for
Quantity
  • Availability
  • Add to Cart