ENA428073149 Enamine

methyl 3,5-dichloro-2-fluorobenzoate

  • MDL number MFCD24108551
  • Cas Number : 1522125-32-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177765

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177765
Availability details for
Quantity
  • Availability
  • Add to Cart