ENA428073151 Enamine

7-bromo-3-chloroquinoline

  • MDL number MFCD18253967
  • Cas Number : 1246549-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177768

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177768
Availability details for
Quantity
  • Availability
  • Add to Cart