ENA428073153 Enamine

(3,5-dibromo-4-chlorophenyl)methanol

  • MDL number MFCD28023767
  • Cas Number : 1795347-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177770

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177770
Availability details for
Quantity
  • Availability
  • Add to Cart