ENA428073154 Enamine

methyl 2-cyano-3,3,3-trifluoropropanoate

  • MDL number MFCD21950345
  • Cas Number : 97821-47-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177772

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177772
Availability details for
Quantity
  • Availability
  • Add to Cart