ENA428073159 Enamine

7H,7aH,12H-isoindolo[2,1-a]perimidin-12-one

  • MDL number MFCD08247257
  • Cas Number : 138937-84-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177780

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177780
Availability details for
Quantity
  • Availability
  • Add to Cart