ENA428073164 Enamine

6-methylpyridine-3-sulfonyl chloride hydrochloride

  • MDL number MFCD26593845
  • Cas Number : 1210868-11-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177795

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177795
Availability details for
Quantity
  • Availability
  • Add to Cart