ENA428073176 Enamine

1-methyl-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-1,3-benzodiazole

  • MDL number MFCD16995902
  • Cas Number : 1107627-02-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177835

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177835
Availability details for
Quantity
  • Availability
  • Add to Cart