ENA428073185 Enamine

1-(2,3-dihydro-1H-isoindol-2-yl)propan-2-one hydrochloride

  • MDL number MFCD28023769
  • Cas Number : 28708-48-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177886

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177886
Availability details for
Quantity
  • Availability
  • Add to Cart