ENA428073186 Enamine

2-(bromomethyl)-5-fluoropyridine hydrobromide

  • MDL number MFCD12546970
  • Cas Number : 1186194-88-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177887

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177887
Availability details for
Quantity
  • Availability
  • Add to Cart