ENA428073187 Enamine

[(2-bromo-4-fluorophenyl)methyl](methyl)amine hydrochloride

  • MDL number MFCD28023645
  • Cas Number : 1645350-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177889

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177889
Availability details for
Quantity
  • Availability
  • Add to Cart