ENA428073194 Enamine

5-methyl-1-benzofuran-2-carbonitrile

  • MDL number MFCD11574554
  • Cas Number : 35351-43-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177926

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177926
Availability details for
Quantity
  • Availability
  • Add to Cart