ENA428073197 Enamine

(4-fluoro-2-methylphenyl)methanesulfonamide

  • MDL number MFCD16686689
  • Cas Number : 1247342-88-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177934

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177934
Availability details for
Quantity
  • Availability
  • Add to Cart