ENA428073198 Enamine

(2,4-dimethylphenyl)methanesulfonamide

  • MDL number MFCD16671939
  • Cas Number : 1078627-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177935

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177935
Availability details for
Quantity
  • Availability
  • Add to Cart