ENA428073199 Enamine

methyl 3-{[(tert-butoxy)carbonyl]amino}-2-hydroxypropanoate

  • MDL number MFCD18447448
  • Cas Number : 113525-87-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177936

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177936
Availability details for
Quantity
  • Availability
  • Add to Cart