ENA428073201 Enamine

(2-aminoethyl)(3,3,3-trifluoropropyl)amine

  • MDL number MFCD18298496
  • Cas Number : 1340301-80-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177940

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177940
Availability details for
Quantity
  • Availability
  • Add to Cart