ENA428073207 Enamine

methyl 6-amino-1,3-dioxaindane-5-carboxylate

  • MDL number MFCD00835593
  • Cas Number : 40680-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177948

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177948
Availability details for
Quantity
  • Availability
  • Add to Cart