ENA428073210 Enamine

3-amino-1-[(4-fluorophenyl)methyl]pyrrolidin-2-one

  • MDL number MFCD14689638
  • Cas Number : 1247800-80-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-177951
Availability details for
Quantity
  • Availability
  • Add to Cart