ENA428073213 Enamine

N-(5-amino-2-methylphenyl)propane-1-sulfonamide hydrochloride

  • MDL number MFCD28023648
  • Cas Number : 1795188-51-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177959

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177959
Availability details for
Quantity
  • Availability
  • Add to Cart