ENA428073220 Enamine

ethyl 4-methylheptanoate

  • MDL number MFCD28023649
  • Cas Number : 22084-69-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177970

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177970
Availability details for
Quantity
  • Availability
  • Add to Cart