ENA428073226 Enamine

1-butyl-5-(propan-2-yl)-1H-pyrazol-4-amine

  • MDL number MFCD21773943
  • Cas Number : 1504703-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177979

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177979
Availability details for
Quantity
  • Availability
  • Add to Cart