ENA428073228 Enamine

2-[2-chloro-6-(pyrrolidin-1-yl)phenyl]ethan-1-amine

  • MDL number MFCD18887168
  • Cas Number : 1338943-69-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177986

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177986
Availability details for
Quantity
  • Availability
  • Add to Cart