ENA428073234 Enamine

N-(2-methoxyethyl)-N-methylpiperidine-3-sulfonamide

  • MDL number MFCD14689194
  • Cas Number : 1249825-79-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178000

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178000
Availability details for
Quantity
  • Availability
  • Add to Cart