ENA428073240 Enamine

1-(4-chlorophenyl)-5-ethyl-1H-pyrazol-4-amine hydrochloride

  • MDL number MFCD28023651
  • Cas Number : 1788596-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-178016

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178016
Availability details for
Quantity
  • Availability
  • Add to Cart