ENA428073241 Enamine

3-amino-N-hydroxypropanamide hydrochloride

  • MDL number MFCD28016697
  • Cas Number : 88872-31-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-142
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-178018

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178018
Availability details for
Quantity
  • Availability
  • Add to Cart