ENA428073246 Enamine

[(3-bromothiophen-2-yl)methyl](2-methoxyethyl)amine

  • MDL number MFCD16093847
  • Cas Number : 1251216-05-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-178034

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178034
Availability details for
Quantity
  • Availability
  • Add to Cart