ENA428073250 Enamine

5-phenyl-1-(propan-2-yl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD13471356
  • Cas Number : 1308236-66-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-178053

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178053
Availability details for
Quantity
  • Availability
  • Add to Cart