ENA428073251 Enamine

1-(2-hydroxyethyl)-5-phenyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD13501679
  • Cas Number : 1292636-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178057

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178057
Availability details for
Quantity
  • Availability
  • Add to Cart