ENA428073258 Enamine

5-ethyl-1-(pentan-3-yl)-1H-pyrazol-4-amine

  • MDL number MFCD21773955
  • Cas Number : 1543192-11-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-178092
Availability details for
Quantity
  • Availability
  • Add to Cart